accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4541-14-4

IUPAC Name: 4-(benzyloxy)butan-1-ol
Synonyms: 4-(benzyloxy)butan-1-ol
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: TYROJDFHUXSBHC-UHFFFAOYSA-N
SMILES: OCCCCOCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)