Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Benzyloxy-1-butanol

Catalog No. 4596905ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-969-05ML 5 mL
45-969-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-969-05ML Supplier Sigma Aldrich Supplier No. 4596905ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-benzyloxy-1-butanol can be prepared starting from butane-1,4-diol.4-benzyloxy-1-butanol may be used for the preparation of ethyl 6-benzyloxy-2-hexenoate, via swern oxidation and wadsworth-emmons olefination
  • It may be used as starting reagent for the synthesis of 4-(benzyloxy)butanal

Specifications

CAS 4541-14-4
Density 1.025 g/mL (at 25°C (literature))
Boiling Point 216°C to 256°C (lit.)
Molecular Formula C11H16O2
Refractive Index n20/D 1.513 (literature)
Linear Formula C6H5CH2O(CH2)4 OH
MDL Number MFCD00155211
Quantity 5 mL
Synonym Tetramethylene glycol monobenzyl ether
Molecular Weight (g/mol) 180.24
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.