accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 4542-47-6

IUPAC Name: 3-(morpholin-4-yl)propanenitrile
Synonyms: 3-(morpholin-4-yl)propanenitrile
Molecular Weight (g/mol): 140.19
Molecular Formula: C7H12N2O
InChi Key: WXVKGHVDWWXBJX-UHFFFAOYSA-N
SMILES: N#CCCN1CCOCC1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)