accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4545-18-0

IUPAC Name: N'-cyclohexylidene-4-methylbenzene-1-sulfonohydrazide
Synonyms: N'-cyclohexylidene-4-methylbenzenesulfonohydrazide
Molecular Weight (g/mol): 266.36
Molecular Formula: C13H18N2O2S
InChi Key: WMCVYCATQIGYIO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NN=C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results