Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone p-Toluenesulfonylhydrazone 98.0+%, TCI America™
SDP

Catalog No. C22955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22955G Supplier TCI America Supplier No. C22955G

Chemical Identifiers

CAS 4545-18-0
Molecular Formula C13H18N2O2S
Molecular Weight (g/mol) 266.36
MDL Number MFCD00161064
InChI Key WMCVYCATQIGYIO-UHFFFAOYSA-N
Synonym Cyclohexanone Tosylhydrazone
PubChem CID 244768
IUPAC Name N'-cyclohexylidene-4-methylbenzene-1-sulfonohydrazide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=C1CCCCC1

Specifications

Quantity 5 g
Formula Weight 266.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexanone p-Toluenesulfonylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.