accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 455-15-2

IUPAC Name: 1-fluoro-4-methanesulfonylbenzene
Synonyms: 1-fluoro-4-methanesulfonylbenzene
Molecular Weight (g/mol): 174.19
Molecular Formula: C7H7FO2S
InChi Key: DPJHZJGAGIWXTD-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results