CAS RN 455-16-3
IUPAC Name:
4-methylbenzene-1-sulfonyl fluoride
Synonyms:
P-fluorosulfonyltoluene
Molecular Weight (g/mol):
174.19
Molecular Formula:
C7H7FO2S
InChi Key:
IZZYABADQVQHLC-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(F)(=O)=O