accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFFF

CAS RN 455-24-3

IUPAC Name: 4-(trifluoromethyl)benzoic acid
Synonyms: 4-trifluoromethylbenzoic acid p-trifluoroformylbenzoic acid P-CF3 benzoic acid α,α,α-trifluoro-p-toluic acid
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: SWKPKONEIZGROQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)