accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOF

CAS RN 455-37-8

IUPAC Name: 3-fluorobenzamide
Synonyms: 3-fluorobenzamide
Molecular Weight (g/mol): 139.13
Molecular Formula: C7H6FNO
InChi Key: YPIGHNIIXYSPKF-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results