Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorobenzamide 97.0+%, TCI America™
SDP

Catalog No. F02665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02665G 5 g
F026625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02665G Supplier TCI America Supplier No. F02665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 455-37-8
Molecular Formula C7H6FNO
Molecular Weight (g/mol) 139.13
MDL Number MFCD00007983
InChI Key YPIGHNIIXYSPKF-UHFFFAOYSA-N
Synonym m-fluorobenzamide, benzamide, 3-fluoro, 3-fluoro-benzamide, benzamide, m-fluoro, 3-fluoranylbenzamide, 3-fluorobenzamide, ksc492m7t, ypighniixyspkf-uhfffaoysa
PubChem CID 68000
IUPAC Name 3-fluorobenzamide
SMILES NC(=O)C1=CC=CC(F)=C1

Specifications

Melting Point 131°C
Color White
Quantity 5 g
Formula Weight 139.13
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.