accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 45521-09-3

IUPAC Name: (4S)-1,3-thiazolidine-4-carboxylic acid
Synonyms: thioproline|(S)-timonacic
Molecular Weight (g/mol): 133.17
Molecular Formula: C4H7NO2S
InChi Key: DZLNHFMRPBPULJ-GSVOUGTGSA-N
SMILES: OC(=O)[C@H]1CSCN1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)