accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHI

CAS RN 45579-91-7

IUPAC Name: 1-phenylmethanamine hydroiodide
Synonyms: benzylamine hydroiodide
Molecular Weight (g/mol): 235.07
Molecular Formula: C7H10IN
InChi Key: PPCHYMCMRUGLHR-UHFFFAOYSA-N
SMILES: I.NCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results