accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4559-79-9

IUPAC Name: [(2E)-4-(dimethylazaniumyl)but-2-en-1-yl]dimethylazanium
Synonyms: [(2E)-4-(dimethylammonio)but-2-en-1-yl]dimethylazanium
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: KUEDAAUECWBMLW-AATRIKPKSA-P
SMILES: C[NH+](C)C\C=C\C[NH+](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results