Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-2-butene-1,4-diamine 97.0+%, TCI America™
SDP

Catalog No. T143610ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T143610ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T143610ML Supplier TCI America Supplier No. T143610ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4559-79-9
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00008336
InChI Key KUEDAAUECWBMLW-AATRIKPKSA-P
Synonym N,N,N′C,N′C-Tetramethyl-1,4-diamino-2-butene
PubChem CID 5365279
IUPAC Name [(2E)-4-(dimethylazaniumyl)but-2-en-1-yl]dimethylazanium
SMILES C[NH+](C)C\C=C\C[NH+](C)C

Specifications

Color Yellow
Quantity 10 mL
UN Number 2734
Formula Weight 142.25
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-2-butene-1,4-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.