accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4559-86-8

IUPAC Name: 1,1,3,3-tetrabutylurea
Synonyms: 1,1,3,3-tetrabutylurea
Molecular Weight (g/mol): 284.49
Molecular Formula: C17H36N2O
InChi Key: SNDGLCYYBKJSOT-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C(=O)N(CCCC)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results