Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,3-Tetrabutylurea 98.0+%, TCI America™
SDP

Catalog No. T25375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25375G 5 g
T253725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25375G Supplier TCI America Supplier No. T25375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4559-86-8
Molecular Formula C17H36N2O
Molecular Weight (g/mol) 284.488
MDL Number MFCD00042901
InChI Key SNDGLCYYBKJSOT-UHFFFAOYSA-N
PubChem CID 20691
IUPAC Name 1,1,3,3-tetrabutylurea
SMILES CCCCN(CCCC)C(=O)N(CCCC)CCCC

Specifications

Melting Point -60°C
Color Yellow
Boiling Point 163°C
Quantity 5 g
Formula Weight 284.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetrabutylurea
RTECSNumber YU2080000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.