accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOCl

CAS RN 456-04-2

IUPAC Name: 2-chloro-1-(4-fluorophenyl)ethan-1-one
Synonyms: 2-chloro-1-(4-fluorophenyl)ethanone
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: UJZWJOQRSMOFMA-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)