accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOOH

CAS RN 456-59-7

IUPAC Name: 3,3,5-trimethylcyclohexyl 2-hydroxy-2-phenylacetate
Synonyms: cyclandelate spasmocyclon ciclospasmol spasmione arto-espasmol cyclergine 环扁桃酯 spasmocyclone sancyclan cyclolyt
Molecular Weight (g/mol): 276.38
Molecular Formula: C17H24O3
InChi Key: WZHCOOQXZCIUNC-UHFFFAOYNA-N
SMILES: CC1CC(CC(C)(C)C1)OC(=O)C(O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results