Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,5-Trimethylcyclohexyl Mandelate (mixture of isomers) 94.0+%, TCI America™
SDP

Catalog No. M082125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M082125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M082125G Supplier TCI America Supplier No. M082125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 456-59-7
Molecular Formula C17H24O3
Molecular Weight (g/mol) 276.38
MDL Number MFCD00056623
InChI Key WZHCOOQXZCIUNC-UHFFFAOYNA-N
Synonym Cyclandelate, Mandelic Acid 3,3,5-Trimethylcyclohexyl Ester
PubChem CID 2893
ChEBI CHEBI:3988
IUPAC Name 3,3,5-trimethylcyclohexyl 2-hydroxy-2-phenylacetate
SMILES CC1CC(CC(C)(C)C1)OC(=O)C(O)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 276.38
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,5-Trimethylcyclohexyl Mandelate (mixture of isomers)
RTECSNumber OO8200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.