accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4584-46-7

IUPAC Name: hydrogen (2-chloroethyl)dimethylamine chloride
Synonyms: hydrogen dimethylaminoethyl chloride chloride
Molecular Weight (g/mol): 144.04
Molecular Formula: C4H11Cl2N
InChi Key: LQLJZSJKRYTKTP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCl
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (1)
  • (3)
  • (4)
  • (1)
  • (7)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)