Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-N,N-dimethylethylamine hydrochloride

Catalog No. D1412085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1412085G 5 g
D141208100G 100 g
D141208500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1412085G Supplier Sigma Aldrich Supplier No. D1412085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-N,N-dimethylethylamine hydrochloride

Specifications

CAS 4584-46-7
Melting Point 201°C to 204°C (lit.)
Molecular Formula C4H10ClN -+ HCl
Linear Formula (CH3)2 NCH2CH2Cl -+ HCl
MDL Number MFCD00012516
Quantity 5 g
Synonym 1-Chloro-2-dimethylaminoethane hydrochloride; 2-(Dimethylamino)ethyl chloride hydrochloride; N-(2-Chloroethyl)dimethylamine hydrochloride
Molecular Weight (g/mol) 144.04
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.