accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NOO(E)

CAS RN 4584-57-0

IUPAC Name: N,N-dimethyl-4-[(1E)-2-(4-nitrophenyl)ethenyl]aniline
Synonyms: N,N-dimethyl-4-[(1E)-2-(4-nitrophenyl)ethenyl]aniline
Molecular Weight (g/mol): 268.32
Molecular Formula: C16H16N2O2
InChi Key: NVLSIZITFJRWPY-ONEGZZNKSA-N
SMILES: CN(C)C1=CC=C(\C=C\C2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results