Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylamino-4'-nitrostilbene 98.0+%, TCI America™
SDP

Catalog No. D25055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25055G Supplier TCI America Supplier No. D25055G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4584-57-0
Molecular Formula C16H16N2O2
Molecular Weight (g/mol) 268.32
MDL Number MFCD00041877
InChI Key NVLSIZITFJRWPY-ONEGZZNKSA-N
Synonym 4-dimethylamino-4'-nitrostilbene, amino-nitrostilbene, dans, n,n-dimethyl-4-2-4-nitrophenyl vinyl aniline, benzenamine, n,n-dimethyl-4-2-4-nitrophenyl ethenyl, 1,4-dimethylamino-4-nitrostilbene, ccris 2642, n,n-dimethyl-4-4-nitrostyryl aniline, e-n,n-dimethyl-4-4-nitrostyryl aniline, n,n-dimethyl-4-2-4-nitrophenyl ethenyl aniline
PubChem CID 5377793
IUPAC Name N,N-dimethyl-4-[(1E)-2-(4-nitrophenyl)ethenyl]aniline
SMILES CN(C)C1=CC=C(\C=C\C2=CC=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Quantity 5 g
UN Number 2811
Formula Weight 268.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylamino-4′-nitrostilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.