accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOH3CCH3CH3

CAS RN 4584-63-8

IUPAC Name: 2,5-bis(2-methylbutan-2-yl)cyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-bis(2-methylbutan-2-yl)cyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 248.37
Molecular Formula: C16H24O2
InChi Key: DHXFOYLEDAOQRR-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results