Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di-tert-amylbenzoquinone 95.0+%, TCI America™
SDP

Catalog No. D21525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21525G Supplier TCI America Supplier No. D21525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4584-63-8
Molecular Formula C16H24O2
Molecular Weight (g/mol) 248.37
MDL Number MFCD00142843
InChI Key DHXFOYLEDAOQRR-UHFFFAOYSA-N
Synonym 2,5-Di-tert-pentyl-1,4-benzoquinone
PubChem CID 97456
IUPAC Name 2,5-bis(2-methylbutan-2-yl)cyclohexa-2,5-diene-1,4-dione
SMILES CCC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)CC

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 248.37
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di-tert-amylbenzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.