accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 459-02-9

IUPAC Name: hydrogen 1-(4-fluorophenyl)propan-2-amine chloride
Synonyms: 4-fluoroamphetamine
Molecular Weight (g/mol): 189.66
Molecular Formula: C9H13ClFN
InChi Key: GKWYMWZWSCKSMT-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(N)CC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results