Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)-2-propylamine, 97%, Thermo Scientific™

Catalog No. AAH51016MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51016MD 250 mg
AAH5101603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51016MD Supplier Thermo Scientific Chemicals Supplier No. H51016MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for drug discovery.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-02-9
Molecular Formula C9H13ClFN
Molecular Weight (g/mol) 189.66
MDL Number MFCD01708101
InChI Key GKWYMWZWSCKSMT-UHFFFAOYNA-N
Synonym 4-fluoroamphetamine, p-fluoroamphetamine, 1-4-fluorophenyl propan-2-amine, 1-4-fluorophenyl propane-2-amine, +--p-fluoroamphetamine, 2-4-fluoro-phenyl-1-methyl-ethylamine, p-fluoro-alpha-methylphenethylamine, 4-fluoro-alpha-methylphenethylamine, benzeneethanamine, p-fluoro-alpha-methyl, 1-4-fluorophenyl-2-propylamine
PubChem CID 9986
IUPAC Name hydrogen 1-(4-fluorophenyl)propan-2-amine chloride
SMILES [H+].[Cl-].CC(N)CC1=CC=C(F)C=C1

Specifications

Quantity 250 mg
Solubility Information Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide.
Formula Weight 153.2
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorophenyl)-2-propylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber SH7040000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.