accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 459-22-3

IUPAC Name: 2-(4-fluorophenyl)acetonitrile
Synonyms: benzeneacetonitrile, 4-fluoro-
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: JHQBLYITVCBGTO-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC#N)C=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)