accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CF

CAS RN 459-47-2

IUPAC Name: 1-ethyl-4-fluorobenzene
Synonyms: 1-ethyl-4-fluorobenzene
Molecular Weight (g/mol): 124.16
Molecular Formula: C8H9F
InChi Key: BLDNWXVISIXWKZ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results