CAS RN 4591-56-4
IUPAC Name:
3,5-diethyl pyridine-3,5-dicarboxylate
Synonyms:
3,5-diethyl pyridine-3,5-dicarboxylate
Molecular Weight (g/mol):
223.23
Molecular Formula:
C11H13NO4
InChi Key:
PGGQCHVPWSXPSI-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(=CN=C1)C(=O)OCC
1
–
1
of
1
results
1
Diethyl 3,5-Pyridinedicarboxylate 98.0+%, TCI America™
CAS: 4591-56-4 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.228 MDL Number: MFCD00085755 InChI Key: PGGQCHVPWSXPSI-UHFFFAOYSA-N Synonym: 3,5-Pyridinedicarboxylic Acid Diethyl Ester PubChem CID: 78340 IUPAC Name: diethyl pyridine-3,5-dicarboxylate SMILES: CCOC(=O)C1=CC(=CN=C1)C(=O)OCC