Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 3,5-Pyridinedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D45965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45965G Supplier TCI America Supplier No. D45965G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4591-56-4
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00085755
InChI Key PGGQCHVPWSXPSI-UHFFFAOYSA-N
Synonym 3,5-Pyridinedicarboxylic Acid Diethyl Ester
PubChem CID 78340
IUPAC Name diethyl pyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=CC(=CN=C1)C(=O)OCC

Specifications

Melting Point 51°C
Boiling Point 117°C
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 3,5-Pyridinedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.