accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBr

CAS RN 460-00-4

IUPAC Name: 1-bromo-4-fluorobenzene
Synonyms: 4-bromofluorobenzene P-bromofluorobenzene
Molecular Weight (g/mol): 175
Molecular Formula: C6H4BrF
InChi Key: AITNMTXHTIIIBB-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (13)
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (9)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (6)
  • (3)
  • (13)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (9)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (6)
  • (5)
  • (1)
  • (1)