Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spex CertiPrep™ GC/MS Tuning Solution
SDP

Catalog No. CLPVTHPK Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLPVTHPK 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLPVTHPK Supplier Spex CertiPrep™ Supplier No. CLPVTHPK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standards for monitoring VOCs by GC/MS

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 524.3, 8260 and CLP Series Methods

Specifications

Chemical Name or Material 4-Bromofluorobenzene
Packaging Bottle
CAS 460-00-4
Concentration 2,500 μg/mL
Color Clear
Melting Point Undetermined
Boiling Point 64.7°C (148.5°F)
Solubility Completely miscible
Solution Type Standard Solution
Type Organic Pollutant Standard
Solvent or Matrix Methanol-P and T
Industry Type Environmental Testing
Quantity 4/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.