accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 461-82-5

IUPAC Name: 4-(trifluoromethoxy)aniline
Synonyms: 4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: XUJFOSLZQITUOI-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC(F)(F)F)C=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)