CAS RN 461432-26-8
IUPAC Name:
2-{4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
dapagliflozin
Molecular Weight (g/mol):
408.88
Molecular Formula:
C21H25ClO6
InChi Key:
JVHXJTBJCFBINQ-UHFFFAOYNA-N
SMILES:
CCOC1=CC=C(CC2=C(Cl)C=CC(=C2)C2OC(CO)C(O)C(O)C2O)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Dapagliflozin
CAS: 461432-26-8 Molecular Formula: C21H25ClO6 Molecular Weight (g/mol): 408.88 InChI Key: JVHXJTBJCFBINQ-UHFFFAOYNA-N IUPAC Name: 2-{4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl}-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: CCOC1=CC=C(CC2=C(Cl)C=CC(=C2)C2OC(CO)C(O)C(O)C2O)C=C1