accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHCO

CAS RN 4616-63-1

IUPAC Name: 2-(prop-2-yn-1-yloxy)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(prop-2-yn-1-yloxy)isoindole-1,3-dione
Molecular Weight (g/mol): 201.18
Molecular Formula: C11H7NO3
InChi Key: HBGZBVPXPDNXOV-UHFFFAOYSA-N
SMILES: O=C1N(OCC#C)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)