Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Propargyloxyphthalimide, 97%, Thermo Scientific™

Catalog No. AAL0923306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0923306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0923306 Supplier Thermo Scientific Chemicals Supplier No. L0923306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4616-63-1
Molecular Formula C11H7NO3
Molecular Weight (g/mol) 201.181
MDL Number MFCD00005890
InChI Key HBGZBVPXPDNXOV-UHFFFAOYSA-N
Synonym n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
PubChem CID 78357
IUPAC Name 2-prop-2-ynoxyisoindole-1,3-dione
SMILES C#CCON1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 150°C to 152°C
Quantity 5 g
Beilstein 1530730
Formula Weight 201.18
Percent Purity 97%
Chemical Name or Material N-Propargyloxyphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.