accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NONHOHO

CAS RN 462-60-2

IUPAC Name: 2-(carbamoylamino)acetic acid
Synonyms: N-(aminocarbonyl)glycine
Molecular Weight (g/mol): 118.09
Molecular Formula: C3H6N2O3
InChi Key: KZVRXPPUJQRGFN-UHFFFAOYSA-N
SMILES: NC(=O)NCC(O)=O
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)