Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydantoic Acid, MP Biomedicals

Catalog No. ICN215598 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215598 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215598 Supplier MP Biomedicals, Inc Supplier No. 0521559801
Add to Cart
Add to Cart

Chemical Identifiers

CAS 462-60-2
Molecular Formula C3H6N2O3
Molecular Weight (g/mol) 118.092
InChI Key KZVRXPPUJQRGFN-UHFFFAOYSA-N
Synonym hydantoic acid, n-carbamoylglycine, carbamoylglycine, glycoluric acid, n-carbamylglycine, glycine, n-aminocarbonyl, acetic acid, ureido, urea, carboxymethyl, 2-ureidoacetic acid, carbomoylglycine
PubChem CID 10020
IUPAC Name 2-(carbamoylamino)acetic acid
SMILES C(C(=O)O)NC(=O)N

Specifications

Quantity 10 g
Formula Weight 118.09
Chemical Name or Material Hydantoic Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.