accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3O

CAS RN 4629-80-5

IUPAC Name: 1,3-dimethylpiperidin-4-one
Synonyms: 1,3-dimethylpiperidin-4-one
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: BGDGMIWDPMJYPP-UHFFFAOYNA-N
SMILES: CC1CN(C)CCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results