Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. D39095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39095G 5 g
D390925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39095G Supplier TCI America Supplier No. D39095G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4629-80-5
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00100893
InChI Key BGDGMIWDPMJYPP-UHFFFAOYSA-N
Synonym 1,3-dimethyl-4-piperidone, 1,3-dimethyl-piperidin-4-one, 4-piperidinone, 1,3-dimethyl, 1,3-dimethyl-4-piperidinone, zlchem 321, pubchem14069, acmc-209k6l, 1,3-dimthylpiperidin-4-one, ksc495i6l, 1,3-dimethylpiperidine-4-one
PubChem CID 107311
IUPAC Name 1,3-dimethylpiperidin-4-one
SMILES CC1CN(CCC1=O)C

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-4-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.