CAS RN 4630-07-3
IUPAC Name:
4a,5-dimethyl-3-(prop-1-en-2-yl)-1,2,3,4,4a,5,6,7-octahydronaphthalene
Synonyms:
valencene
Molecular Weight (g/mol):
204.36
Molecular Formula:
C15H24
InChi Key:
QEBNYNLSCGVZOH-UHFFFAOYNA-N
SMILES:
CC1CCC=C2CCC(CC12C)C(C)=C
1
–
1
of
1
results
1
Sigma Aldrich (+)-Valencene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.