Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Valencene

Catalog No. 7505650GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-056-50GF 50 g
75-056-10GF 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-056-50GF Supplier Sigma Aldrich Supplier No. 7505650GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-Valencene is a sesquiterpene, which contains decalin framework and two methyl-bearing stereocenters. It is an aroma component of citrus fruit.(+)-Valencene can be used as a precursor to prepare: (+)-Nootkatone (a sesquiterpene) by dark singlet oxygenation. Benzoyloxyvalencene by reacting with tert-butyl peroxy benzoate via Kharasch-Sosnovsky allylic oxidation method. (+)-Lineariifolianone, a natural product.

Specifications

CAS 4630-07-3
Density 0.92 g/mL at 25°C
Boiling Point 274°C
Molecular Formula C15H24
Linear Formula C15H24
Refractive Index n20/D 1.504
Quantity 50 g
MDL Number MFCD00075884
Synonym (3R,4aS,5R)-4a,5-Dimethyl-3-isopropenyl-1,2,3,4,4a,5,6,7-octahydronaphthalene
Molecular Weight (g/mol) 204.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.