CAS RN 4630-20-0
CAS RN 4630-20-0
IUPAC Name:
3-methyl-9H-carbazole
Synonyms:
3-methylcarbazole
Molecular Weight (g/mol):
181.24
Molecular Formula:
C13H11N
InChi Key:
PHKYYUQQYARDIU-UHFFFAOYSA-N
SMILES:
CC1=CC2=C(NC3=C2C=CC=C3)C=C1
1
–
1
of
1
results
1
3-Methyl-9H-carbazole 98.0+%, TCI America™
CAS: 4630-20-0 Molecular Formula: C13H11N Molecular Weight (g/mol): 181.238 MDL Number: MFCD00464338 InChI Key: PHKYYUQQYARDIU-UHFFFAOYSA-N PubChem CID: 20746 IUPAC Name: 3-methyl-9H-carbazole SMILES: CC1=CC2=C(C=C1)NC3=CC=CC=C32