Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. M25611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25611G Supplier TCI America Supplier No. M25611G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4630-20-0
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00464338
InChI Key PHKYYUQQYARDIU-UHFFFAOYSA-N
PubChem CID 20746
IUPAC Name 3-methyl-9H-carbazole
SMILES CC1=CC2=C(C=C1)NC3=CC=CC=C32

Specifications

Melting Point 208°C
Color White-Yellow
Boiling Point 365°C
Quantity 1 g
Formula Weight 181.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.