accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 46331-50-4

IUPAC Name: 5-methoxybenzene-1,3-dicarboxylic acid
Synonyms: 5-methoxybenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 196.16
Molecular Formula: C9H8O5
InChi Key: POSMIIJADZKUPL-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(=C1)C(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results