Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxyisophthalic Acid 98.0+%, TCI America™
SDP

Catalog No. M18355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18355G Supplier TCI America Supplier No. M18355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 46331-50-4
Molecular Formula C9H8O5
Molecular Weight (g/mol) 196.158
MDL Number MFCD00272364
InChI Key POSMIIJADZKUPL-UHFFFAOYSA-N
PubChem CID 327428
IUPAC Name 5-methoxybenzene-1,3-dicarboxylic acid
SMILES COC1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 270°C
Color White
Quantity 5 g
Formula Weight 196.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyisophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.