accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHO

CAS RN 4634-09-7

IUPAC Name: (6-phenylpyridin-3-yl)methanol
Synonyms: (6-phenylpyridin-3-yl)methanol
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: RXXIVABOKBNIEI-UHFFFAOYSA-N
SMILES: OCC1=CN=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results