accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N(E)

CAS RN 4635-87-4

IUPAC Name: (3E)-pent-3-enenitrile
Synonyms: (3E)-pent-3-enenitrile
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: UVKXJAUUKPDDNW-NSCUHMNNSA-N
SMILES: C\C=C\CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results