Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pentenenitrile, TCI America™
SDP

Catalog No. P095825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P095825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P095825ML Supplier TCI America Supplier No. P095825ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4635-87-4
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00001963
InChI Key UVKXJAUUKPDDNW-NSCUHMNNSA-N
Synonym 2-Buten-1-yl Cyanide, 1-Cyano-2-butene
PubChem CID 5324707
IUPAC Name (E)-pent-3-enenitrile
SMILES CC=CCC#N

Specifications

Color Yellow
Boiling Point 144°C
UN Number 3275
Quantity 25 mL
Formula Weight 81.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pentenenitrile
EINECSNumber (2)-3154
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.